Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM8A Rabbit anti-Human, Mouse, Rat, Clone: 5K5E6, Novus Biologicals™
SDP

Catalog No. NB166705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166705 100 μg
NB166706 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166705 Supplier Novus Biologicals Supplier No. NBP315805100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RBM8A Monoclonal antibody specifically detects RBM8A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen RBM8A
Applications Western Blot
Classification Monoclonal
Clone 5K5E6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Binder of OVCA1-1, BOV-1, BOV-1A, BOV-1B, BOV-1C, MDS014, RBM8BRBM8, ribonucleoprotein RBM8, Ribonucleoprotein RBM8A, RNA binding motif protein 8A, RNA binding motif protein 8B, RNA-binding motif protein 8A, RNA-binding protein 8A, RNA-binding protein Y14, Y14, ZNRP, ZRNP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RBM8A (Q9Y5S9).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9939
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.