Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBMS1 Antibody (OTI2H1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273834V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273834V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273834V Supplier Novus Biologicals Supplier No. NBP273834V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBMS1 Monoclonal specifically detects RBMS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RBMS1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H1
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chromosome 2 open reading frame 12, DKFZp564H0764, HCC-4, MGC15146, MGC3331, MGC97258, MGC97270, MGC97282, MGC99543, MSSP1, MSSP-1, MSSP-2, MSSP-3, RNA binding motif, single stranded interacting protein 1, SCR2MGC70597, single-stranded-interacting protein 1, suppressor of cdc 2 (cdc13) with RNA binding motif 2
Gene Symbols RBMS1
Host Species Mouse
Immunogen Full length recombinant protein of human RBMS1 (NP_058520) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5937.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.