Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBMS2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen RBMS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB5727120UL
SDP
View Documents
Novus Biologicals
NBP15727120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP157271
SDP
View Documents
Novus Biologicals
NBP157271
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

RBMS2 Polyclonal specifically detects RBMS2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RBMS2
Polyclonal
Rabbit
Q15434
5939
Synthetic peptides corresponding to RBMS2(RNA binding motif, single stranded interacting protein 2) The peptide sequence was selected from the N terminal of RBMS2. Peptide sequence MLLSVTSRPGISTFGYNRNNKKPYVSLAQQMAPPSPSNSTPNSSSGSNGN.
Primary
Western Blot
Unconjugated
RUO
FLJ39093, FLJ40023, RNA binding motif, single stranded interacting protein 2, RNA-binding motif, single-stranded-interacting protein 2, SCR3FLJ43262, Suppressor of CDC2 with RNA-binding motif 3
RBMS2
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.