Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBMS2 Antibody (PCRP-RBMS2-1B6), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB187711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187711 Supplier Novus Biologicals Supplier No. NBP314163DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBMS2 Monoclonal antibody specifically detects RBMS2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen RBMS2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-RBMS2-1B6
Conjugate DyLight 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ39093, FLJ40023, RNA binding motif, single stranded interacting protein 2, RNA-binding motif, single-stranded-interacting protein 2, SCR3FLJ43262, Suppressor of CDC2 with RNA-binding motif 3
Host Species Mouse
Immunogen Recombinant full-length human RBMS2 protein (Uniprot: Q15434)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5939
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.