Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP4/Retinol-Binding Protein 4 Antibody (393005) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB372402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372402 Supplier Novus Biologicals Supplier No. FAB3378L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 393005
Conjugate DyLight 550
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RBP4/Retinol-Binding Protein 4, Glu19-Leu201, Accession # P02753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.