Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP4/Retinol-Binding Protein 4 Antibody (423619) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB372309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372309 Supplier Novus Biologicals Supplier No. FAB34761W
Add to Cart
Add to Cart

Rat Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 423619
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4, Met1-Leu201, Accession # NP_035385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5950
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.