Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP4/Retinol-Binding Protein 4 Antibody (RB42), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB358991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358991 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358991 Supplier Novus Biologicals Supplier No. NBP105426AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Western Blot, ELISA
Classification Monoclonal
Clone RB42
Conjugate Alexa Fluor 750
Dilution Western Blot, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human recombinant RBP4.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5950
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.