Learn More
RBP4/Retinol-BindingProtein4 Rabbit anti-Human, Clone: JA33-10, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | RBP4/Retinol-BindingProtein4 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | JA33-10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin |
| Formulation | TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide |
| Gene Alias | interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma |
| Gene Symbols | RBP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.