Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP5 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB080692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080692 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080692 Supplier Novus Biologicals Supplier No. NBP297021R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBP5 Polyclonal specifically detects RBP5 in Human samples. It is validated for ELISA.

Specifications

Antigen RBP5
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias Cellular retinol-binding protein III, CRBP3, CRBPIII, CRBP-III, HRBPiso, putative cellular retinol-binding protein CRBP III, retinol binding protein 5, cellular, retinol-binding protein 5, retinol-binding protein 5, cellular
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RBP5 (P82980; Met1-Arg135).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83758
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.