Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPJ Monoclonal Antibody (1F1), Active Motif™

Rat Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | RBPJ |
---|---|
Clone | 1F1 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3460
![]() ![]() |
Active Motif
61506 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3459
![]() ![]() |
Active Motif
61505 |
100 μg |
Each for $638.65
|
|
|||||
Description
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.Specifications
RBPJ | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
AI843960; AOS3; BOS_6494; CBF1; CBF-1; csl; H-2K binding factor-2; IGKJRB; IGKJRB1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; LOC100346517; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; Recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; Renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog | |
RBPJ | |
IgG2b | |
Protein G | |
Antibody |
1F1 | |
ChIP Assay, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rat | |
Human, Mouse | |
P31266, Q06330 | |
19664, 3516 | |
This antibody was raised against a full-length recombinant protein corresponding to human RBPJ. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
RBPJ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title