Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAN1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP146853Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146853Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146853Z Supplier Novus Biologicals Supplier No. NBP146853AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RCAN1 Polyclonal specifically detects RCAN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RCAN1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P53805
Gene Alias ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1
Gene Symbols RCAN1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot# P53805]
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1827
Test Specificity Human and mouse. Immunogen has 92% identity to rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.