Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAN3 Antibody (OTI1B2), Novus Biologicals™
SDP

Catalog No. NBP245928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245928 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245928 Supplier Novus Biologicals Supplier No. NBP245928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunofluorescence

RCAN3 Monoclonal specifically detects RCAN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.

Specifications

Antigen RCAN3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI1B2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunofluorescence 1:100
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias Down syndrome candidate region 1-like 2, Down syndrome critical region gene 1-like 2, DSCR1L2regulator of calcineurin 3 isoform 13,45,MCIP3regulator of calcineurin 3 isoform 1a2,34,5, hRCN3, Myocyte-enriched calcineurin-interacting protein 3, RCAN family member 3, RCN3, Regulator of calcineurin 3, regulator of calcineurin 3 isoform 1a3,45,Down syndrome candidate region 1-like protein 2, regulator of calcineurin 3 isoform 1b2,34,5, regulator of calcineurin 3 isoform 1c2,34,5, regulator of calcineurin 3 isoform 1c3,45,calcipressin-3
Gene Symbols RCAN3
Host Species Mouse
Immunogen Full length human recombinant protein of human RCAN3(NP_038469) produced in E.coli.
Molecular Weight of Antigen 27.3 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11123
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.