Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCC1 Antibody (3D11), Novus Biologicals™
SDP

Catalog No. NB018810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018810 0.025 mg
NBP197543 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018810 Supplier Novus Biologicals Supplier No. NBP1975430.025MG

Mouse Monoclonal Antibody

RCC1 Monoclonal specifically detects RCC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen RCC1
Applications Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone 3D11
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 2 ug/ml
Formulation PBS (pH 7.2) and 1.0% Sucrose with No Preservative
Gene Alias Cell cycle regulatory protein, CHC1RCC1-I, chromosome condensation 1, Chromosome condensation protein 1, guanine nucleotide-releasing protein, regulator of chromosome condensation, regulator of chromosome condensation 1, SNHG3-RCC1, SNHG3-RCC1 readthrough transcript
Gene Symbols RCC1
Host Species Mouse
Immunogen Recombinant human RanGEF.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1104
Test Specificity Recognizes Human, Mouse and Rat RanGEF.
Reconstitution Reconstitute with 0.1 ml sterilized distilled water to a final concentration of 1.0 mg/ml.
Target Species Human, Rat, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.