Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCC1 Antibody (3D11), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $548.00
Specifications
Antigen | RCC1 |
---|---|
Clone | 3D11 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 2 ug/ml |
Applications | Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
Description
RCC1 Monoclonal specifically detects RCC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
RCC1 | |
LYOPH | |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
Cell cycle regulatory protein, CHC1RCC1-I, chromosome condensation 1, Chromosome condensation protein 1, guanine nucleotide-releasing protein, regulator of chromosome condensation, regulator of chromosome condensation 1, SNHG3-RCC1, SNHG3-RCC1 readthrough transcript | |
RCC1 | |
IgG1 | |
Protein A purified | |
Store at -20C. Avoid freeze-thaw cycles. |
3D11 | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunoprecipitation 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Mouse | |
1104 | |
Recombinant human RanGEF. | |
Primary | |
Reconstitute with 0.1 ml sterilized distilled water to a final concentration of 1.0 mg/ml. | |
Recognizes Human, Mouse and Rat RanGEF. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title