Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107237
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5107237 100 μL
1 options

Catalog No. PIPA5107237

Supplier: Invitrogen™ PA5107237

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

Probable molecular chaperone assisting protein biosynthesis and transport in the endoplasmic reticulum (PubMed:16433634, PubMed:28939891). Required for the proper biosynthesis and transport of pulmonary surfactant-associated protein A/SP-A, pulmonary surfactant-associated protein D/SP-D and the lipid transporter ABCA3. By regulating both the proper expression and the degradation through the endoplasmic reticulum-associated protein degradation pathway of these proteins plays a crucial role in pulmonary surfactant homeostasis. Has an anti-fibrotic activity by negatively regulating the secretion of type I and type III collagens (PubMed:28939891). This calcium-binding protein also transiently associates with immature PCSK6 and regulates its secretion (PubMed:16433634). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCN3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene RCN3
Gene Accession No. Q96D15
Gene Alias 6030455P07Rik; D530026G20Rik; D7Ertd671e; EF-hand calcium-binding protein RLP49; Rcn3; reticulocabin; reticulocalbin 3; reticulocalbin 3, EF-hand calcium binding domain; reticulocalbin-3; RLP49; UNQ239/PRO272
Gene Symbols RCN3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human RCN3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.