Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH11 Antibody (OTI1B4), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273843A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273843A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273843A405 Supplier Novus Biologicals Supplier No. NBP273843AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RDH11 Monoclonal specifically detects RDH11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RDH11
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B4
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Androgen-regulated short-chain dehydrogenase/reductase 1, ARSDR1CGI82, EC 1.1.1.300, HCBP12, HCV core-binding protein HCBP12, MDT1, prostate short-chain dehydrogenase reductase 1, Prostate short-chain dehydrogenase/reductase 1, PSDR1FLJ32633, RALR1, Retinal reductase 1, retinol dehydrogenase 11, retinol dehydrogenase 11 (all-trans and 9-cis), retinol dehydrogenase 11 (all-trans/9-cis/11-cis), SCALD, SDR7C1, short chain dehydrogenase/reductase family 7C, member 1
Gene Symbols RDH11
Host Species Mouse
Immunogen Full length recombinant protein of human RDH11 (NP_057110) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 51109.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.