Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP158017

 View more versions of this product

Catalog No. NBP158017


Only null left
Add to Cart

Description

Description

RDH12 Polyclonal specifically detects RDH12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RDH12
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
All-trans and 9-cis retinol dehydrogenase, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.100, FLJ30273, LCA13retinol dehydrogenase 12, all-trans and 9-cis, LCA3, retinol dehydrogenase 12, retinol dehydrogenase 12 (all-trans and 9-cis), retinol dehydrogenase 12 (all-trans/9-cis/11-cis), SDR7C2, short chain dehydrogenase/reductase family 7C, member 2
Rabbit
Affinity purified
RUO
Primary
Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Zebrafish: 92%; Bovine: 84%; Chicken: 84%; Pig: 84%; Rabbit: 84%; Guinea pig: 76%.
Human, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Q96NR8
RDH12
Synthetic peptides corresponding to RDH12(retinol dehydrogenase 12 (all-trans/9-cis/11-cis)) The peptide sequence was selected from the middle region of RDH12. Peptide sequence AKRLQGTGVTTYAVHPGVVRSELVRHSSLLCLLWRLFSPFVKTAREGAQT.
100 μL
Vision
145226
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.