Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH14 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB135360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135360 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135360 Supplier Novus Biologicals Supplier No. NBP306302AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RDH14 Polyclonal specifically detects RDH14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RDH14
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Alcohol dehydrogenase PAN2, EC 1.1.1.-, PAN2retinol dehydrogenase 14, pancreas protein 2, retinol dehydrogenase 14 (all-trans and 9-cis), retinol dehydrogenase 14 (all-trans/9-cis/11-cis), SDR7C4, short chain dehydrogenase/reductase family 7C, member 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RDH14 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57665
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.