Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ready-Made Luria Broth (LB) Powder
Click to view available options
Quantity:
1 kg
Description
- Off-white free flowing powder
- pH(2.5%, H2O): 7.0 ± 0.2
Contents
- Casein Peptone: 40%
- Yeast Extract: 20%
- Sodium Chloride: 40%

Specifications
Specifications
Color | Off-white |
Form | Powder |
Product Type | Microbiology Media |
For Use With (Application) | Propagation of Bacteria |
Packaging Type | Plastic Bottle |
Quantity | 1 kg |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction