Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Ready-Made Terrific Broth Powder
Click to view available options
Quantity:
1 kg
Description
- Off-white free flowing powder
- pH(5.08%, H2O): 7.2 ± 0.2
Contents
- Casein Peptone: 12g/L
- Yeast Extract: 24g/L
- Potassium Phosphate Dibasic: 12.5g/L
- Potassium Phosphate Monobasic: 2.3g/L

Specifications
Specifications
Color | Off-white |
Form | Powder |
Product Type | Microbiology Media |
For Use With (Application) | Provides Greater Yields of Plasmid DNA and Recombinant Proteins |
Packaging Type | Plastic Bottle |
Quantity | 1 kg |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction