Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Reboxetine mesylate
GREENER_CHOICESDP

Catalog No. 198250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-825-0 50 mg
19-821-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-825-0 Supplier Tocris Bioscience™ Supplier No. 1982/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective noradrenalin uptake inhibitor; orally active

Potent and selective inhibitor of noradrenalin uptake (Ki values are 1.1, 129 and > 10000 nM for rat NET, SERT and DAT respectively). Displays > 1000-fold selectivity over α-adrenoceptors, 5-HT, dopamine and muscarinic ACh receptors. Orally active antidepressant.

Chemical Identifiers

CAS 98769-84-7
Molecular Formula C20H27NO6S

Specifications

Quantity 50 mg
Formula Weight 409.5
Purity >99%
Molecular Weight (g/mol) 409.497
Chemical Name or Material Reboxetine mesylate
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.