Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REC8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP187145 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP187145 0.1 mL
NB406203 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP187145 Supplier Novus Biologicals Supplier No. NBP187145

Rabbit Polyclonal Antibody

REC8 Polyclonal specifically detects REC8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen REC8
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias Cohesin Rec8p, cohesion rec8p, HR21spB, meiotic recombination and sister chromatid cohesion phosphoprotein of therad21p family, meiotic recombination protein REC8 homolog, meiotic recombination protein REC8-like 1, MGC950, REC8 homolog (yeast), REC8L1human homolog of rad21, S. pombe, REC8-like 1, REC8-like 1 (yeast), Rec8p, recombination and sister chromatid cohesion protein homolog
Gene Symbols REC8
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PSPFDIPQIRHLLEAAIPERVEEIPPEVPTEPREPERIPVTVLPPEAITILEAEPIRMLEIEGERELPEVSRRELDLLI
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9985
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.