Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RecA-1 Antibody (HIS52), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1064647A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064647A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064647A32 Supplier Novus Biologicals Supplier No. NB10064647AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RecA-1 Monoclonal specifically detects RecA-1 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen RecA-1
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIS52
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias RecA protein, RecA1, RecA2, recaLP, Recombinase A
Host Species Mouse
Immunogen Stromal cells from rat lymph node
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells.
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.