Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Canine IL-31 Protein
SDP

Catalog No. p-7183005 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
250 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a un-tagged and corresponding to the amino acids 24-159aa of Canine IL31.

Source: Baculovirus

Amino Acid Sequence:

SHMAPTHQLP PSDVRKIILE LQPLSRGLLE DYQKKETGVP ESNRTLLLCL TSDSQPPRLN SSAILPYFRA IRPLSDKNII DKIIEQLDKL KFQHEPETEI SVPADTFECK SFILTILQQF SACLESVFKS LNSGPQ

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 20% glycerol
Gene ID (Entrez) 386653
Molecular Weight (g/mol) 15.3 kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias IL-31interleukin-31, interleukin 31
Gene Symbol IL31
Buffer PBS (pH 7.4), 20% glycerol.
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.