Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Human alpha-Synuclein Active, Filaments Protein
SDP

Catalog No. p-7229464 Shop All Novus Biologicals Products
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

For Use With (Application) Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS pH 7.4
Gene ID (Entrez) 6622
Quantity 100 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Gene Symbol SNCA
Research Category Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Protein alpha-Synuclein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.