Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human alpha-Synuclein Oligomers, (Dopamine HCL Stabilized) Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Specifications
Specifications
For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
Formulation | PBS pH 7.4 |
Gene ID (Entrez) | 6622 |
Quantity | 100 μg |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Gene Alias | alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein |
Gene Symbol | SNCA |
Research Category | Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience |
Purity or Quality Grade | >95%, by SDS-PAGE |
Protein | alpha-Synuclein |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction