Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human BDNF Protein
SDP

Catalog No. NB002057 Shop All Novus Biologicals Products
Click to view available options
Quantity:
50 μg

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein for Human BDNF The Recombinant Human BDNF Protein is derived from E. coli. The Recombinant Human BDNF Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Sterile filtered and lyophilized with no additives.
Gene ID (Entrez) 627
Molecular Weight (g/mol) 27.0 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Gene Symbol BDNF
Reconstitution Reconstitute in H2O to a concentration of 0.1 - 1 μg/μL. The solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Sterile filtered and lyophilized with no additives.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.