Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD34 Protein
SDP

Catalog No. p-7120954 Shop All Novus Biologicals Products
Click to view available options
Quantity:
250 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 32-290 of Human CD34 The Recombinant Human CD34 Protein is derived from Baculovirus. The Recombinant Human CD34 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 947
Molecular Weight (g/mol) 28.6kDa
Name Human CD34 Protein
Purification Method >95%, by PAGE
Quantity 50 μg
Source Baculovirus
Immunogen ADLSLDNNGT ATPELPTQGT FSNVSTNVSY QETTTPSTLG STSLHPVSQH GNEATTNITE TTVKFTSTSV ITSVYGNTNS SVQSQTSVIS TVFTTPANVS TPETTLKPSL SPGNVSDLST TSTSLATSPT KPYTSSSPIL SDIKAEIKCS GIREVKLTQG ICLEQNKTSS CAEFKKDRGE GLARVLCGEE QADADAGAQV CSLLLAQSEV RPQCLLLVLA NRTEISSKLQ LMKKHQSDLK KLGILDFTEQ DVASHQSYSQ KTHHHHHH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Gene Symbol CD34
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.