Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human CLEC4E His Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 41-219 of Human CLEC4E The Recombinant Human CLEC4E Protein is derived from E. coli. The Recombinant Human CLEC4E Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 0.25mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | 20mM Tris 8.0 containing 10% glycerol |
Gene ID (Entrez) | 26253 |
Molecular Weight (g/mol) | TMW: 22.9kDa |
Quantity | 0.1 mg |
Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
Gene Alias | CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 |
Common Name | CLEC4E |
Conjugate | Unconjugated |
Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction