Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FGF-21 Protein
SDP

Catalog No. NB002058 Shop All Novus Biologicals Products
Click to view available options
Quantity:
100 μg

Highly purified. Generating reliable and reproducible results.

Recombinant protein containing 182 amino acids for Human FGF-21 The Recombinant Human FGF-21 Protein is derived from E. coli. The Recombinant Human FGF-21 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Formulation Lyopohilized from 10 mM Tris (pH 7.5), 100 mM NaCl.
Gene ID (Entrez) 26291
Molecular Weight (g/mol) 19.5 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias FGF-21, fibroblast growth factor 21
Gene Symbol FGF21
Reconstitution Reconstitute with sterile H2O to a concentration of 0.1 mg/mL, which can be further diluted into other aqueous solutions.
Buffer Lyopohilized from 10 mM Tris (pH 7.5), 100 mM NaCl.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.