Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FGF-22 Protein
SDP

Catalog No. p-7161308 Shop All Novus Biologicals Products
Click to view available options
Quantity:
100 μg
500 μg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

An un-tagged bioactive recombinant protein corresponding to Human FGF-22

The Recombinant Human FGF-22 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 27006
Molecular Weight (g/mol) 17.3 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/ug protein (determined by Kinetic LAL Method).
Gene Symbol FGF22
Specific Reactivity FGF-22
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.