Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human FGF-6 Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
1 mg
25 μg
5 μg
Description
An un-tagged bioactive recombinant protein corresponding to Human FGF-6
The Recombinant Human FGF-6 Protein has been validated for the following applications: SDS-Page, Bioactivity.
The Recombinant Human FGF-6 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Gene ID (Entrez) | 2251 |
| Molecular Weight (g/mol) | 18.9 kDa |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 5 μg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | FGF6 |
| Specific Reactivity | FGF-6 is a monomer, 18.9 kDa (169 aa) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction