Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Glutathione Peroxidase 3/GPX3 FLAG Protein
SDP

Catalog No. p-7249155 Shop All R&D Systems Products
Click to view available options
Quantity:
10 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number P22352
Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Liquid. 0.2um-filtered solution in PBS, pH 7.2
Gene ID (Entrez) 2878
Molecular Weight (g/mol) M.W. Theoretical: 30 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias EC 1.11.1, EC 1.11.1.9, Extracellular glutathione peroxidase, glutathione peroxidase 3, glutathione peroxidase 3 (plasma), GPx-3, GPx-P, GPXP, GSHPx-3, GSHPx-P, Plasma glutathione peroxidase
Gene Symbol GPX3
Research Category Cancer, Cell Biology, Lipid and Metabolism, Signal Transduction
Purity or Quality Grade >90% SDS-PAGE
Protein Glutathione Peroxidase 3/GPX3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.