Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human Glycine Receptor Alpha 1 GST (N-Term) Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
2 μg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 2741 |
| Molecular Weight (g/mol) | 78.1 kDa |
| Quantity | 2 μg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | GLRA 1, Glycine receptor 48 kDa subunit, Glycine receptor strychnine-binding subunit, glycine receptor subunit alpha-1, glycine receptor, alpha 1, glycine receptor, alpha 1 (startle disease/hyperekplexia), MGC138878, MGC138879, STHE |
| Gene Symbol | GLRA1 |
| Research Category | Diabetes Research, Neuronal Cell Markers, Neuroscience, Neurotransmission |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction