Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-1 alpha/IL-1F1 Protein
SDP

Catalog No. p-7249176 Shop All R&D Systems Products
Click to view available options
Quantity:
100 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

D10.G4.1 cell proliferation, >2.0 x 10^7 units/mg (ED50 acceptance criteria are <50 pg/mL).

Specifications

For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from a 0.2 micron filtered solution, 10 mM sodium phosphate, pH 7.5
Gene ID (Entrez) 3552
Quantity 100 μg
Storage Requirements Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method).
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Gene Symbol IL1A
Reconstitution Sterile water at 0.1 mg/mL
Research Category Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Purity or Quality Grade >95%, by SDS-PAGE
Protein IL-1 alpha/IL-1F1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.