Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-10R alpha His Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.05 mg
0.25 mg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-235 of Human IL-10 R alpha The Recombinant Human IL-10R alpha Protein is derived from Baculovirus. The Recombinant Human IL-10R alpha Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.2mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 3587 |
| Molecular Weight (g/mol) | TMW: 25.2kDa |
| Quantity | 0.05 mg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha |
| Common Name | IL-10 R alpha |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction