Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-15 R alpha Protein
SDP

Catalog No. p-7145906 Shop All R&D Systems Products
Click to view available options
Quantity:
250 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page, Cell Proliferation

A bioactive recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 31-205 of Human IL-15 R alpha The Recombinant Human IL-15R alpha Protein is derived from Baculovirus. The Recombinant Human IL-15R alpha Protein has been validated for the following applications: SDS-Page, Cell Proliferation.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% Glycerol, 1mM DTT
Gene ID (Entrez) 3601
Molecular Weight (g/mol) 45.6 kDa
Quantity 250 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol IL15RA
Cross Reactivity IL-15 R alpha
Species Human
Purity or Quality Grade >80%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.