Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Kallikrein 10 His Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-276 of Human Kallikrein 10 The Recombinant Human Kallikrein 10 Protein is derived from E. coli. The Recombinant Human Kallikrein 10 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol 0.4M UREA |
| Gene ID (Entrez) | 5655 |
| Molecular Weight (g/mol) | TMW: 29.1kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | breast normal epithelial cell associated serine protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.74, Kallikrein 10, kallikrein-related peptidase 10, NES1kallikrein-10, Normal epithelial cell-specific 1, Protease serine-like 1, PRSSL1protease, serine-like, 1 |
| Common Name | Kallikrein 10 |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction