Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human OR4D2 GST (N-Term) Protein
SDP

Catalog No. NB170842 Shop All R&D Systems Products
Click to view available options
Quantity:
2 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) ELISA, Immunoaffinity Purification, Protein Array, Western Blot
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 124538
Molecular Weight (g/mol) M.W. Theoretical: 60.72 kDa
Quantity 2 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias BC2009, B-lymphocyte membrane protein BC2009, G-protein coupled membrane protein, olfactory receptor 4D2, olfactory receptor OR17-24, olfactory receptor, family 4, subfamily D, member 2, OR17-24
Gene Symbol OR4D2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein OR4D2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.