Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human p70 S6 Kinase/S6K Isoform a Protein
SDP

Catalog No. p-7145930 Shop All Novus Biologicals Products
Click to view available options
Quantity:
10 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-525 of Human p70 S6 Kinase/S6K The Recombinant Human p70 S6 Kinase/S6K Isoform a Protein is derived from Baculovirus. The Recombinant Human p70 S6 Kinase/S6K Isoform a Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation 50mM Tris-HCl (pH 7.5) containing 30% glycerol, 0.5M NaCl, 2mM DTT, 0.1mM PMSF
Gene ID (Entrez) 6198
Molecular Weight (g/mol) 60.2 kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol RPS6KB1
Cross Reactivity p70 S6 Kinase/S6K
Species Human
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.