Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human POP7 His Protein
SDP

Catalog No. NBP25176101 Shop All R&D Systems Products
Click to view available options
Quantity:
0.02 mg
0.1 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-140 of Human POP7 The Recombinant Human POP7 Protein is derived from E. coli. The Recombinant Human POP7 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 50% glycerol, 2mM DTT, 2mM EDTA
Gene ID (Entrez) 10248
Molecular Weight (g/mol) TMW: 18.3kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias 0610037N12Rik, EC 3.1.26.5, hPOP7, processing of precursor 7, ribonuclease P subunit, processing of precursor 7, ribonuclease P subunit (S. cerevisiae), processing of precursor 7, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p20, Ribonucleases P/MRP protein subunit POP7 homolog, RNaseP protein p20, RPP20S. cerevisiae) homolog
Common Name POP7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.