Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human SCN3B His Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
250 μg
50 μg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-159 of Human SCN3B The Recombinant Human SCN3B Protein is derived from Baculovirus. The Recombinant Human SCN3B Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. PBS (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 55800 |
Molecular Weight (g/mol) | 16.8 kDa |
Name | Recombinant Human SCN3B Protein |
Purification Method | >90%, by SDS-PAGE |
Quantity | 50 μg |
Source | Baculovirus |
Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction