Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human SDH Assembly Factor 1 His Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.02 mg
0.1 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-115 of Human SDH Assembly Factor 1 The Recombinant Human SDH Assembly Factor 1 Protein is derived from E. coli. The Recombinant Human SDH Assembly Factor 1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.25mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH7.4), 30% glycerol, 2mM DTT, 0.1mM PMSF |
| Gene ID (Entrez) | 644096 |
| Molecular Weight (g/mol) | TMW: 15.2kDa |
| Quantity | 0.02 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | LYR Motif Containing 8, LYR Motif-Containing Protein 8, LYRM8, SDHAF1, Succinate Dehydrogenase Assembly Factor 1, Mitochondrial, Succinate Dehydrogenase Complex Assembly Factor 1 |
| Common Name | SDH Assembly Factor 1 |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction