Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Skp1 Protein
SDP

Catalog No. p-7237472 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.1 mg
500 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to the amino acids 1- 160 of Human Skp1 The Recombinant Human Skp1 Protein is derived from E. coli. The Recombinant Human Skp1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_008861
For Use With (Application) ELISA, Peptide ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 50mM NaCl, 10% glycerol
Gene ID (Entrez) 6500
Molecular Weight (g/mol) 18kDa
Name Skp1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1- 160aa, Human
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.