Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TGF-beta 3 Animal-Free Protein
SDP

Catalog No. N276236100U Shop All R&D Systems Products
Click to view available options
Quantity:
100 μg
500 μg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

An un-tagged bioactive recombinant protein corresponding to Human TGF-beta 3

The Recombinant Human TGF-beta 3 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 7043
Molecular Weight (g/mol) 25.7 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C.
Regulatory Status RUO
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/ug protein (determined by Kinetic LAL Method).
Gene Symbol TGFB3
Specific Reactivity TGF-beta 3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 10 mM acetic acid and 20% Ethanol
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.