Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein
SDP

Catalog No. p-7145941 Shop All R&D Systems Products
Click to view available options
Quantity:
250 μg
50 μg

Catalog No. NB261985250

Supplier: Novus Biologicals™ NBP261985250UG

Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal hIgG-His-tag and corresponding to the amino acids 27-126 of Human TGF-beta RI/ALK-5 The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein is derived from Baculovirus. The Recombinant Human TGF-beta RI/ALK-5 Isoform 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 7046
Molecular Weight (g/mol) 38 kDa
Quantity 250 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol TGFBR1
Cross Reactivity TGF-beta RI/ALK-5
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.