Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TRANCE/TNFSF11/RANK L Animal-Free Protein
SDP

Catalog No. NB27619010U Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 μg
2 μg

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • An un-tagged bioactive recombinant protein corresponding to Human TRANCE/TNFSF11/RANK L
  • Amino Acid Sequence: EKAMVDGSW LDLAKRSKLE AQPFAHLTIN ATDIPSGSHK VSLSSWYHDR GWAKISNMTF SNGKLIVNQD GFYYLYANIC FRHHETSGDL ATEYLQLMVY VTKTSIKIPS SHTLMKGGST KYWSGNSEFH FYSINVGGFF KLRSGEEISI EVSNPSLLDP DQDATYFGAF KVRDID
  • Produced with no materials of animal or human origin using ANIMAL-FREE purification processes
  • No animal-derived ingredients, No FBS
  • E. coli based production system
  • Animal-Free facility, fermentor, columns, filters, labware, lyophilization, and storage
  • Endotoxin levels at <1 EUs/μg
  • Purity levels at 95% or greater

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 19.7 kDa
Purification Method >95%, by SDS-PAGE
Quantity 10 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol TNFSF11
Specific Reactivity TRANCE/TNFSF11/RANK L is a monomer, 19.7 kDa (175 aa)
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.