Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ULBP-1 Protein
SDP

Catalog No. p-7120984 Shop All R&D Systems Products
Click to view available options
Quantity:
250 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 26-216 of Human ULBP-1 The Recombinant Human ULBP-1 Protein is derived from Baculovirus. The Recombinant Human ULBP-1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 80329
Molecular Weight (g/mol) 23.4kDa
Name Human ULBP-1 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Immunogen ADLGWVDTHC LCYDFIITPK SRPEPQWCEV QGLVDERPFL HYDCVNHKAK AFASLGKKVN VTKTWEEQTE TLRDVVDFLK GQLLDIQVEN LIPIEPLTLQ ARMSCEHEAH GHGRGSWQFL FNGQKFLLFD SNNRKWTALH PGAKKMTEKW EKNRDVTMFF QKISLGDCKM WLEEFLMYWE QMLDPTKPPS LAPGHHHHHH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1
Gene Symbol ULBP1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.