Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human WISP2 Protein
SDP

Catalog No. p-7180298 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
20 μg
5 μg

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to Human WISP2

The Recombinant Human WISP2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 8839
Molecular Weight (g/mol) 24.4 kDa
Purification Method >95%, by SDS-PAGE
Quantity 5 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol WISP2
Specific Reactivity WISP2 is a monomer, 24.4 kDa (228 aa)
Reconstitution Sterile 10 mM acetic acid at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.