Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse HGFR/c-MET hIgG-His Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.05 mg
Specifications
Specifications
Concentration | 0.25 mg/mL |
For Use With (Application) | SDS-PAGE |
Gene ID (Entrez) | 4233 |
Quantity | 0.05 mg |
Source | Baculovirus |
Storage Requirements | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles |
Endotoxin Concentration | < 1.0 EU per 1μg of protein (determined by LAL method) |
Species | Mouse |
Buffer | PBS (pH 7.4), 10% glycerol |
Purity or Quality Grade | > 85%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction